missing translation for 'onlineSavingsMsg'
Learn More

Urea, Honeywell Fluka™

Product Code. 16645743 Shop All Honeywell Fluka Products
Change view
Click to view available options
2.5KG POLY BOTTLE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16645743 2.5KG POLY BOTTLE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16645743 Supplier Honeywell Fluka Supplier No. 15604H2.5KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 57-13-6
Physical Form Crystalline Granules
Quantity 2.5 kg
Molecular Formula CH4N2O
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILES NC(N)=O
IUPAC Name urea
Molecular Weight (g/mol) 60.06
PubChem CID 1176
ChEBI CHEBI:48376
Percent Purity 99.0 to 100.5%
Loss on Drying Max. 1.0 %
Packaging Plastic Bottle
Appearance Complying
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.