missing translation for 'onlineSavingsMsg'
Learn More

Urea, Extra Pure, SLR, Fisher Chemical™

Product Code. 10132740 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
3 kg
Unit Size:
1kg
3kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10132740 500 g 500g
10142740 1 kg 1kg
10716311 3 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10132740 Supplier Fisher Chemical Supplier No. U/0450/53
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number 8022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Melting Point 131°C
Color White
pH 7.5 to 9.5
Packaging Plastic powder jar
Quantity 500 g
Formula Weight 60.06g/mol
Vapor Pressure 1.25mmHg 25
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.