missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Urea-d4, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Urea-d4 |
| CAS | 1433-11-0 |
| Molecular Formula | C D4 N2 O |
| Quantity | 10 g |
| Synonym | Urea-d4 (6CI,7CI,8CI,9CI), 1,1,3,3-Tetradeuteriourea, Urea-d4, Urea-D4, Carbamimidic acid-d4, Carbamimidic-d4 acid, Carbonyl diamide-d4, Urea D4, [2H4]-Urea |
| InChI Formula | InChI=1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)/i/hD4 |
| SMILES | [2H]N([2H])C(=O)N([2H])[2H] |
| IUPAC Name | 1,1,3,3-tetradeuteriourea |
| Molecular Weight (g/mol) | 64.08 |
| Formula Weight | 64.057 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction