missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Urea (Colorless-to-White Crystals or Crystalline Powder/Mol. Biol.), Fisher BioReagents™
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Poly Bottle
Poly Drum
Poly Pail
Description
Urea is used as a denaturing agent for DNA and proteins.
Specifications
Absorbance | 0.05 max. (5M solution) at 280nm |
Melting Point | 131°C |
Color | White |
pH | 7.5 to 9.5 |
Assay Percent Range | ≥99 % |
Packaging | Poly Bottle |
Linear Formula | NH2CONH2 |
Quantity | 500 g |
Merck Index | 15, 10052 |
Formula Weight | 60.06 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'