missing translation for 'onlineSavingsMsg'
Learn More

Urea, Certified AR for Analysis, Fisher Chemical™

Product Code. p-7073651 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
3 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
3kg
500g
5kg
This item is not returnable. View return policy

Product Code. 10102790

Brand: Fisher Chemical U/0500/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CH4N2O, CAS Number-57-13-6, carbamimidic acid, alphadrate, carbonyldiamide, isourea, urevert, ureaphil, pseudourea, ureophil, carbonyldiamine, carbamide, 5kg, CHEBI:48376, White, 60.06g/mol, XSQUKJJJFZCRTK-UHFFFAOYSA-N, 131 deg.C, 8022, 60.056, Plastic keg, 7.5 to 9.5, Solid, 1176, C(-O)(N)N

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number 8022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Melting Point 131°C
Color White
pH 7.5 to 9.5
Packaging Plastic powder jar
Quantity 500 g
Formula Weight 60.06g/mol
Vapor Pressure 1.25mmHg 25
Physical Form Solid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.