missing translation for 'onlineSavingsMsg'
Learn More

Urea, 99%, (White crystalline powder or pellets/Ultra Pure), MP Biomedicals

Product Code. 11412002 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 lb.
5 lb.
Unit Size:
1lb
5lb
This item is not returnable. View return policy

Product Code. 11412002

Brand: MP Biomedicals 0210569501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Color White
Assay Percent Range 99%
Quantity 1 lb.
Formula Weight 60.1
Grade Ultrapure
Physical Form Crystalline Powder or Pellets
Assay 4X Crystallized
Chemical Name or Material Urea
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.