missing translation for 'onlineSavingsMsg'
Learn More

Urea, 99%, ACS reagent

Product Code. 10135413 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
10kg
2.5kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10135413 2.5 kg Plastic bottle 2.5kg
10462985 100 g Plastic bottle 100g
10200001 500 g Plastic bottle 500g
11936921 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10135413 Supplier Thermo Scientific Acros Supplier No. 424580025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 57-13-6 | CH4N2O | 60.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-13-6
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Decomposition Information >132°C
Melting Point 131.0°C to 135.0°C
Color White
Ignition Residue 0.01% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Molecular Formula CH4N2O
Linear Formula H2NCONH2
Quantity 2.5 kg
Beilstein 03, 42
Fieser 01,1262; 02,455
Merck Index 15, 10052
Solubility Information Solubility in water: 1080g/L (20°C). Other solubilities: soluble in alcohol,benzene,concentrated hcl,practically insoluble in chloroform and ether,soluble in methanol: 16.6g/100 mL,soluble in ethanol: 5g/100 mL
Molecular Weight (g/mol) 60.06
Formula Weight 60.06
Vapor Pressure 1.25mmHg at 25°C
Percent Purity 99%
Grade ACS Reagent
Insoluble Matter 0.01% max
Physical Form Fine Crystals
Chemical Name or Material Urea
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.