missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Urea, 99.3+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Description
Urea has important use as a fertilizer and feed supplement. Used for the preparation of plastics and drugs. Urea and its derivatives plays an important role in the cosmetics industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 132°C to 135°C |
Color | White |
Density | 1.335 g/mL |
pH | 9.2 to 9.5 |
Odor | Ammonia-like |
Linear Formula | NH2CONH2 |
Quantity | 100 g |
Beilstein | 635724 |
Merck Index | 14,9867 |
Solubility Information | Soluble in water,glycerol and ethanol. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'