missing translation for 'onlineSavingsMsg'
Learn More

Urea, ≥98%, Molecular Biology Reagent Grade, MP Biomedicals™

Product Code. 11476641 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11476641 5 kg 5kg
11446641 100 g 100g
11456641 500 g 500g
11466641 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11476641 Supplier MP Biomedicals Supplier No. 0219485794
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.056
MDL Number MFCD00008022
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
PubChem CID 1176
ChEBI CHEBI:48376
IUPAC Name urea
SMILES C(=O)(N)N

Specifications

Color White
Density 1.335g/mL at 25°C (Lit.)
Assay Percent Range ≥98%
Quantity 5 kg
Formula Weight 60.06
Grade Molecular Biology Reagent
Physical Form Powder
Chemical Name or Material Urea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.