missing translation for 'onlineSavingsMsg'
Learn More

Urapidil Hydrochloride, TRC

Product Code. 30008138
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008138 500 mg 500mg
17973085 100 mg 100mg
17982225 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008138 Supplier Toronto Research Chemicals Supplier No. TRCU815400500MG
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64887-14-5
Molecular Formula C20 H29 N5 O3 . Cl H
Molecular Weight (g/mol) 423.94
Synonym 2,4(1H,3H)-Pyrimidinedione, 6-[[3-[4-(2-methoxyphenyl)-1-piperazinyl]propyl]amino]-1,3-dimethyl-, hydrochloride (1:1), 2,4(1H,3H)-Pyrimidinedione, 6-[[3-[4-(2-methoxyphenyl)-1-piperazinyl]propyl]amino]-1,3-dimethyl-, monohydrochloride (9CI), Urapidil hydrochloride
IUPAC Name 6-[3-[4-(2-methoxyphenyl)piperazin-1-yl]propylamino]-1,3-dimethylpyrimidine-2,4-dione;hydrochloride
SMILES Cl.COc1ccccc1N2CCN(CCCNC3=CC(=O)N(C)C(=O)N3C)CC2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C20H29N5O3.ClH/c1-22-18(15-19(26)23(2)20(22)27)21-9-6-10-24-11-13-25(14-12-24)16-7-4-5-8-17(16)28-3;/h4-5,7-8,15,21H,6,9-14H2,1-3H3;1H
Formula Weight 423.2037
Percent Purity >95%
Chemical Name or Material Urapidil Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.