missing translation for 'onlineSavingsMsg'
Learn More

Urapidil Hydrochloride, TRC

Product Code. 17982225
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17982225 50 mg 50mg
30008138 500 mg 500mg
17973085 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17982225 Supplier Toronto Research Chemicals Leverandørnr. TRCU81540050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 64887-14-5
Molecular Formula C20 H29 N5 O3 . Cl H
Molecular Weight (g/mol) 423.94
Synonym 2,4(1H,3H)-Pyrimidinedione, 6-[[3-[4-(2-methoxyphenyl)-1-piperazinyl]propyl]amino]-1,3-dimethyl-, hydrochloride (1:1), 2,4(1H,3H)-Pyrimidinedione, 6-[[3-[4-(2-methoxyphenyl)-1-piperazinyl]propyl]amino]-1,3-dimethyl-, monohydrochloride (9CI), Urapidil hydrochloride
IUPAC Name 6-[3-[4-(2-methoxyphenyl)piperazin-1-yl]propylamino]-1,3-dimethylpyrimidine-2,4-dione;hydrochloride
SMILES Cl.COc1ccccc1N2CCN(CCCNC3=CC(=O)N(C)C(=O)N3C)CC2

Tekniske data

Quantity 50 mg
InChI Formula InChI=1S/C20H29N5O3.ClH/c1-22-18(15-19(26)23(2)20(22)27)21-9-6-10-24-11-13-25(14-12-24)16-7-4-5-8-17(16)28-3;/h4-5,7-8,15,21H,6,9-14H2,1-3H3;1H
Formula Weight 423.2037
Percent Purity >95%
Chemical Name or Material Urapidil Hydrochloride
Grade HPLC
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.