missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Urapidil hydrochloride

Product Code. 17231216 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17231216 5 g 5g
17221216 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17231216 Supplier Thermo Scientific Chemicals Supplier No. 467590050
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 64887-14-5 | C20H30ClN5O3, C20H29N5O3·HCl | 423.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64887-14-5
Molecular Formula C20H30ClN5O3

Specifications

Quantity 5 g
Melting Point 249.0°C to 250.0°C
Formula Weight 423.94
InChI Key KTMLZVUAXJERAT-UHFFFAOYSA-N
SMILES [H+].[Cl-].COC1=CC=CC=C1N1CCN(CCCNC2=CC(=O)N(C)C(=O)N2C)CC1
IUPAC Name hydrogen 6-({3-[4-(2-methoxyphenyl)piperazin-1-yl]propyl}amino)-1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione chloride
Molecular Weight (g/mol) 423.94
Chemical Name or Material Urapidil hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.