missing translation for 'onlineSavingsMsg'
Learn More

Uranium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

Catalog Number p-5533099
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-61-1, 7697-37-2, 7732-18-5
U
238.029
JFALSRSLKYAFGM-UHFFFAOYSA-N
23989
CHEBI:49936
uranium
[U]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-61-1, 7697-37-2, 7732-18-5
U
238.029
JFALSRSLKYAFGM-UHFFFAOYSA-N
23989
CHEBI:49936
uranium
[U]

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
U1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

CAS Min % 0.1, 2.0, 97.9
CAS Max % 0.1, 2.0, 97.9
Grade Assurance
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLU2-2Y, -2X    10,000    PLU2-3Y, -3X
Packaging Triple-leached LDPE bottle
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
Solvent or Matrix 2% HNO3
Quantity 125 mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Chemical Name or Material Uranium Standard (Depleted)
Show More Show Less
compliance-icons
Product Identifier
  • 1000
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : nitric acid, Uraniumfrom Uranyl NitrateHexahydrate

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'