missing translation for 'onlineSavingsMsg'
Learn More

Uracil-5-carboxaldehyde, 97%

Product Code. 15497977
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497977 1 g 1g
15407987 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497977 Supplier Thermo Scientific Alfa Aesar Supplier No. H51098.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1195-08-0 | C5H4N2O3 | 140.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1195-08-0
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.10
MDL Number MFCD00192185
InChI Key OHAMXGZMZZWRCA-UHFFFAOYSA-N
Synonym 5-formyluracil, 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbaldehyde, uracil 5-carbaldehyde, uracil-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 1,2,3,4-tetrahydro-2,4-dioxo, 2,4 1h,3h-pyrimidinedione, 5-formyl, 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxaldehyde, 2,3h-pyrimidinedione, 5-formyl, 5-pyrimidinecarboxaldehyde,2,3,4-tetrahydro-2,4-dioxo, zlchem 870
PubChem CID 304590
ChEBI CHEBI:80961
IUPAC Name 2,4-dioxo-1H-pyrimidine-5-carbaldehyde
SMILES O=CC1=CNC(=O)NC1=O

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 140.1
Percent Purity 97%
Chemical Name or Material Uracil-5-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.