Learn More
RETSCH UR 3 Ultrasonic Bath
Clean test sieves and grinding tools quickly and easily. Retsch™ UR 3 Ultrasonic Bathes are designed for the simultaneous cleaning of up to 5 test sieves up to 203mm in diameter.
Brand:  RETSCH 70.791.0006
Additional Details : Weight : 27.50000kg
 View more versions of this product
Product Code. 11964762
  • £7595.00 / Each
Estimated Shipment: 09-07-2024
to see stock.
Add to basket
Description
Description
  • Gentle yet thorough cleaning of test sieves in an ultrasonic bath increases their working lives
  • Damage which could occur during manual cleaning is avoided
  • Fast, gentle, and highly efficient cleaning
  • Universal and compact
  • Easy to use, saves time and cost
  • Intensive dispersion and degassing
  • Low maintenance
  • Long life
Specifications
Specifications
Yes
110/130 V
30 cm
Cleaning test sieves and grinding tools quickly and easily
UR 3
50 cm
50/60 Hz
45 L
110 to 130 V, 50/60 Hz
30 cm
50 cm