missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UQCRQ Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106446
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total UQCRQ.
This gene encodes a ubiquinone-binding protein of low molecular mass. This protein is a small core-associated protein and a subunit of ubiquinol-cytochrome c reductase complex III, which is part of the mitochondrial respiratory chain.
Specifications
| UQCRQ | |
| Polyclonal | |
| Unconjugated | |
| UQCRQ | |
| 1100001F06Rik; 1500040F11Rik; 5830407L17Rik; 9.5kDa; AA959903; c1502; Complex III subunit 8; complex III subunit VII; complex III subunit VIII; Cytochrome b-c1 complex subunit 8; low molecular mass ubiquinone-binding protein; low molecular mass ubiquinone-binding protein (9.5kD); low molecular mass ubiquinone-binding protein (Qpc-pending); MC3DN4; QCR8; QPC; QP-C; transcript under zygotic control; ubiquinol-cytochrome c reductase binding protein; ubiquinol-cytochrome c reductase complex 9.5 kDa protein; ubiquinol-cytochrome c reductase complex III subunit VII; ubiquinol-cytochrome c reductase complex ubiquinone-binding protein QP-C; ubiquinol-cytochrome c reductase, complex III subunit VII; ubiquinol-cytochrome c reductase, complex III subunit VII, 9.5kDa; unknown; UQCR7; Uqcrb; UQCRQ | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22272, 27089, 497902 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14949, Q7TQ16, Q9CQ69 | |
| UQCRQ | |
| A synthesized peptide derived from human UQCRQ(Accession O14949), corresponding to amino acid residues M1-R48. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction