missing translation for 'onlineSavingsMsg'
Learn More

UQCRQ Polyclonal Antibody
GREENER_CHOICE

UQCRQ Polyclonal Antibody

Brand:  Thermo Scientific PA561285

Product Code. 15779101

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

UQCRQ Polyclonal Antibody for ICC/IF, IHC

TRUSTED_SUSTAINABILITY
Specifications

Specifications

UQCRQ
Polyclonal
Unconjugated
UQCRQ
1100001F06Rik; 1500040F11Rik; 5830407L17Rik; 9.5kDa; AA959903; c1502; Complex III subunit 8; complex III subunit VII; complex III subunit VIII; Cytochrome b-c1 complex subunit 8; low molecular mass ubiquinone-binding protein; low molecular mass ubiquinone-binding protein (9.5kD); low molecular mass ubiquinone-binding protein (Qpc-pending); MC3DN4; QCR8; QPC; QP-C; transcript under zygotic control; ubiquinol-cytochrome c reductase binding protein; ubiquinol-cytochrome c reductase complex 9.5 kDa protein; ubiquinol-cytochrome c reductase complex III subunit VII; ubiquinol-cytochrome c reductase complex ubiquinone-binding protein QP-C; ubiquinol-cytochrome c reductase, complex III subunit VII; ubiquinol-cytochrome c reductase, complex III subunit VII, 9.5kDa; unknown; UQCR7; Uqcrb; UQCRQ
Rabbit
Antigen affinity chromatography
RUO
27089
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O14949
UQCRQ
Recombinant protein corresponding to Human UQCRQ. Recombinant protein control fragment (Product #RP-100764).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.