missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ UQCRQ Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30630
This item is not returnable.
View return policy
Description
UQCRQ Polyclonal antibody specifically detects UQCRQ in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| UQCRQ | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| complex III subunit 8, cytochrome b-c1 complex subunit 8, low molecular mass ubiquinone-binding protein (9.5kD), QPC, ubiquinol-cytochrome c reductase complex ubiquinone-binding protein QP-C, ubiquinol-cytochrome c reductase, complex III subunit VII, 9.5kDa, UQCR7 | |
| Between 32 and 82 | |
| 100 μg | |
| Cell Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 27089 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction