missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UQCRQ Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£328.00 - £513.00
Specifications
| Antigen | UQCRQ |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18412861
|
Novus Biologicals
NBP1-92563-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18055717
|
Novus Biologicals
NBP1-92563 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UQCRQ Polyclonal specifically detects UQCRQ in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| UQCRQ | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 27089 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:REFGNLTRMRHVISYSLSPFEQRAYPHVFTKGIPNVLRR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| complex III subunit 8, cytochrome b-c1 complex subunit 8, low molecular mass ubiquinone-binding protein (9.5kD), QPC, ubiquinol-cytochrome c reductase complex ubiquinone-binding protein QP-C, ubiquinol-cytochrome c reductase, complex III subunit VII, 9.5kDa, UQCR7 | |
| UQCRQ | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title