missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UQCRFS1 Recombinant Rabbit Monoclonal Antibody (24GB2460)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557830
This item is not returnable.
View return policy
Description
UQCRFS1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is a respiratory chain that generates an electrochemical potential coupled to ATP synthesis.
Specifications
| UQCRFS1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Uqcrfs1 | |
| 4430402G14Rik; 8 kDa subunit 9; AI875505; Complex III subunit 5; Complex III subunit IX; Cytochrome b-c1 complex subunit 11; Cytochrome b-c1 complex subunit 5; Cytochrome b-c1 complex subunit 9; Cytochrome b-c1 complex subunit Rieske, mitochondrial; liver regeneration-related protein LRRGT00195; LRRGT00195; Rieske iron-sulfur protein; Rieske protein UQCRFS1; RIP1; RIS1; RISP; Su9; Ubiquinol-cytochrome c reductase 8 kDa protein; Ubiquinol-cytochrome c reductase iron-sulfur subunit; ubiquinol-cytochrome c reductase, Rieske iron-sulfur polypeptide 1; UQCR5; UQCRFS1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 291103, 66694, 7386 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2460 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P20788, P47985, Q9CR68 | |
| Uqcrfs1 | |
| A synthesized peptide derived from human UQCRFS1 (aa 220-274). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction