missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UQCRFS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18443-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to UQCRFS1.
Component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is a respiratory chain that generates an electrochemical potential coupled to ATP synthesis.Specifications
| UQCRFS1 | |
| Polyclonal | |
| Unconjugated | |
| UQCRFS1 | |
| Complex III subunit 5, Complex III subunit IX, Rieske iron sulfur protein, RIP1, RIS1, RISP, UQCR5, UQCRFS1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 291103, 66694, 7386 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P20788, P47985, Q9CR68 | |
| Uqcrfs1 | |
| UQCRFS1 Fusion Protein Ag13315 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction