missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: RVKKPEWVIL IGVCTHLGCV PIANAGDFGG YYCPCHGSHY DASGRIRLGP APLNLEVPTY EFTSDDMVIV Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
Component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is a respiratory chain that generates an electrochemical potential coupled to ATP synthesis.
Specifications
Specifications
| Antigen | UQCRFS1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Uqcrfs1 |
| Gene Accession No. | P20788, P47985 |
| Gene Alias | 4430402G14Rik; 8 kDa subunit 9; AI875505; Complex III subunit 5; Complex III subunit IX; Cytochrome b-c1 complex subunit 11; Cytochrome b-c1 complex subunit 5; Cytochrome b-c1 complex subunit 9; Cytochrome b-c1 complex subunit Rieske, mitochondrial; liver regeneration-related protein LRRGT00195; LRRGT00195; Rieske iron-sulfur protein; Rieske protein UQCRFS1; RIP1; RIS1; RISP; Su9; Ubiquinol-cytochrome c reductase 8 kDa protein; Ubiquinol-cytochrome c reductase iron-sulfur subunit; ubiquinol-cytochrome c reductase, Rieske iron-sulfur polypeptide 1; UQCR5; UQCRFS1 |
| Gene Symbols | Uqcrfs1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
