Learn More
Invitrogen™ UQCRC2 Polyclonal Antibody

Description
Immunogen sequence: GVSHPVLKQV AEQFLNMRGG LGLSGAKANY RGGEIREQNG DSLVHAAFVA ESAVAGSAEA NAFSVLQHVL GAGPHVKRGS NTTSHLHQAV AKATQQPFDV SAFNASYSDS GLFGIYTISQ ATAAGDVIKA AYNQVKTIAQ GNLSNTDVQA AKNKLKAGYL MSVESSECFL EEVGSQALVA GSYMPPSTVL QQIDSVANAD IINAAKKFVS GQKSMAASGN LGHTPFVDEL; Positive Samples: U-251MG, MCF7, HepG2, H460, Mouse kidney, Mouse brain, Mouse liver, Rat spinal cord, Rat testis, NCI-H460; Cellular Location: Matrix side, Mitochondrion inner membrane, Peripheral membrane protein.
Specifications
Specifications
| Antigen | UQCRC2 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.76 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | UQCRC2 |
| Gene Accession No. | P22695, P32551, Q9DB77 |
| Gene Alias | 1500004O06Rik; complex III subunit 2; Core protein II; cytochrome bc-1 complex core protein II; cytochrome b-c1 complex subunit 2, mitochondrial; MC3DN5; QCR2; U MC3DN5; ubiquinol cytochrome c reductase core protein 2; ubiquinol-cytochrome C reductase complex core protein 2, mitochondrial precursor (Complex III subunit II); ubiquinol-cytochrome c reductase core protein II; ubiquinol-cytochrome-c reductase complex core protein 2; UQCR2; Uqcrc2 |
| Gene Symbols | UQCRC2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.