missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UQCRB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£143.00 - £251.00
Specifications
| Antigen | UQCRB |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of lysate, Immunohistochemistry-Paraffin 1:000-1:10000 |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18661678
|
Novus Biologicals
NBP3-12711-100ul |
100 μg |
£251.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18651497
|
Novus Biologicals
NBP3-12711-50ul |
50 μg |
£143.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UQCRB Polyclonal antibody specifically detects UQCRB in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| UQCRB | |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology, Cell Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| PBS, pH7.0 | |
| 7381 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of lysate, Immunohistochemistry-Paraffin 1:000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Complex III subunit 7, Complex III subunit VII, cytochrome b-c1 complex subunit 7, QCR7, QPC, QP-CFLJ92016, ubiquinol-cytochrome c reductase binding protein, Ubiquinol-cytochrome c reductase complex 14 kDa protein, ubiquinol-cytochrome c reductase, complex III subunit VI, ubiquinone-binding protein, UQBC, UQBPFLJ97033, UQCR6, UQPC | |
| Produced in rabbits immunized with E. coli-derived Human UQCRB fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title