missing translation for 'onlineSavingsMsg'
Learn More

UQCR10 Antibody, Novus Biologicals™

Product Code. 18266203 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18266203 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18266203 Supplier Novus Biologicals Supplier No. NBP162349

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UQCR10 Polyclonal specifically detects UQCR10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UQCR10
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9UDW1
Gene Alias Complex III subunit 9, Complex III subunit X, cytochrome b-c1 complex subunit 9, Cytochrome c1 non-heme 7 kDa protein, cytochrome C1, nonheme 7kDa protein, HSPC051, HSPC151, QCR9, ubiquinol-cytochrome c reductase complex (7.2 kD), ubiquinol-cytochrome c reductase, complex III subunit X, ubiquinol-cytochrome c reductase, complex III subunit X, 7.2kDa, UCCR7.2, UCRCUbiquinol-cytochrome c reductase complex 7.2 kDa protein
Gene Symbols UQCR10
Host Species Rabbit
Immunogen Synthetic peptides corresponding to UCRC(ubiquinol-cytochrome c reductase complex (7.2 kD)) The peptide sequence was selected from the middle region of UCRC. Peptide sequence LFRRTSTFALTIIVGVMFFERAFDQGADAIYDHINEGKLWKHIKHKYENK.
Molecular Weight of Antigen 7 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29796
Test Specificity This product is specific to Subunit or Isoform: 9.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.