Learn More
Invitrogen™ UQCC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114251
Description
Immunogen sequence: RCQMPDTFNS WFLITLLHVW MCLVRMKQEG RSGKYMCRII VHFMWEDVQQ For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
Heme oxygenase, an essential enzyme in heme catabolism, cleaves heme to form biliverdin, which is subsequently converted to bilirubin by biliverdin reductase, and carbon monoxide, a putative neurotransmitter. Heme oxygenase activity is induced by its substrate heme and by various nonheme substances. Heme oxygenase occurs as 2 isozymes, an inducible heme oxygenase-1 and a constitutive heme oxygenase-2. HMOX1 and HMOX2 belong to the heme oxygenase family.
Specifications
| UQCC1 | |
| Polyclonal | |
| Unconjugated | |
| UQCC1 | |
| 2310079L17Rik; 2410003P15Rik; 3110038N19Rik; basic FGF repressed, Zic binding protein; basic FGF repressed, zinc binding protein; basic FGF-repressed Zic-binding protein; bFGF-repressed Zic-binding protein; BFZB; Bfzp; BZFB; C20orf44; Cbp3; cytochrome B protein synthesis 3 homolog; FLJ10850; mbFZb; ubiquinol-cytochrome c reductase complex assembly factor 1; ubiquinol-cytochrome c reductase complex chaperone; ubiquinol-cytochrome c reductase complex chaperone CBP3 homolog; ubiquinol-cytochrome c reductase complex chaperone, CBP3 homolog; ubiquinol-cytochrome-c reductase complex assembly factor 1; UQCC; UQCC1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55245 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NVA1 | |
| UQCC1 | |
| Synthetic peptide directed towards the N-terminal region of Human UQCC (aa 136-185). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.