missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UPRT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531684
This item is not returnable.
View return policy
Description
Recommended positive controls: A549. Predicted reactivity: Mouse (98%), Zebrafish (90%), Pig (100%), Chicken (100%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes uracil phosphoribosyltransferase, which catalyzes the conversion of uracil and 5-phosphoribosyl-1-R-diphosphate to uridine monophosphate (UMP). This reaction is an important part of nucleotide metabolism, specifically the pyrimidine salvage pathway. The enzyme localizes to the nucleus and cytoplasm. The protein is a potential target for rational design of drugs to treat parasitic infections and cancer.
Specifications
| UPRT | |
| Polyclonal | |
| Unconjugated | |
| UPRT | |
| FUR1; Gm774; RP11-311P8.3; UMP pyrophosphorylase; UPP; Uprt; UPRTase; uracil phosphoribosyltransferase (FUR1) homolog; uracil phosphoribosyltransferase (FUR1) homolog (S. cerevisiae); uracil phosphoribosyltransferase homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 139596, 331487 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| B1AVZ0, Q96BW1 | |
| UPRT | |
| Recombinant protein encompassing a sequence within the center region of human UPRT. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction