missing translation for 'onlineSavingsMsg'
Learn More

Uprosertib, MedChemExpress

Product Code. 16447016
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16447016 100 mg 100mg
16446938 10 mg 10mg
16422997 50 mg 50mg
16409487 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16447016 Supplier Medchem Express Supplier No. HY15965
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Uprosertib (GSK2141795) is a potent and selective pan-Akt inhibitor with IC50 values of 180/328/38 nM for Akt1/Akt2/Akt3, respectively.

Uprosertib (GSK2141795) is a potent and selective pan-Akt inhibitor with IC50 values of 180/328/38 nM for Akt1/Akt2/Akt3, respectively.

Specifications

Chemical Name or Material Uprosertib
CAS 1047634-65-0
Color White
Purity Grade Notes Research
Molecular Formula C18H16Cl2F2N4O2
Quantity 100 mg
Synonym GSK2141795
Solubility Information DMSO : 50 mg/mL (116.48 mM; Need ultrasonic)
SMILES O=C(C1=CC(C2=C(Cl)C=NN2C)=C(Cl)O1)N[C@@H](CC3=CC=C(F)C(F)=C3)CN
Molecular Weight (g/mol) 429.25
Formula Weight 429.25
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.