missing translation for 'onlineSavingsMsg'
Learn More

Manutan Upright frame for rapid pallet racking, 3500mm x 900mm

Product Code. 12876228
Encompass_Preferred
This item is not returnable. View return policy

Product Code. 12876228

Brand: Manutan A363768

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use to replace damaged frames or to build bespoke pallet racking. Heavy duty steel uprights, fitted with heavy duty base plates. Diagonals and horizontals galvanised steel. Hard-wearing powder coated finish in blue. Manufactured to European standards of FEM and UK standard BS EN ISO 9001 2000.

  • Blue
  • Steel
  • Dimensions (H x D): 3500mm x 900mm
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.