missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Upright frame for rapid pallet racking, 2500mm x 1100mm
Description
Use to replace damaged frames or to build bespoke pallet racking. Heavy duty steel uprights, fitted with heavy duty base plates. Diagonals and horizontals galvanised steel. Hard-wearing powder coated finish in blue. Manufactured to European standards of FEM and UK standard BS EN ISO 9001 2000.
- Blue
- Steel
- Dimensions (H x D): 2500mm x 1100mm
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?