missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation UPLC Method Development Scouting Kit

Product Code. p-7236731
Encompass
Click to view available options
Length (Metric):
50 mm
100 mm
Diameter (Metric):
2.1 mm
3 mm
Unit Size:
Pack of 4
This item is not returnable. View return policy

Product Code. 13149979

Brand: Waters Corporation 176001603

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The UPLC method development scouting kit combines BEH C18, Shield RP18, Phenyl and HSS T3 column chemistries to accomodate your method development approach, enabling methods to be developed efficiently and effectively.

  • By utilizing UPLC Technology for method development, a substantial improvement in throughput can be realized.
  • This, in turn, reduces cost per sample and time of analysis considerably while maintaining or improving separation integrity, enabling products to be brought to market faster and improving the overall profitability of the assay.

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 50 mm
Max. Pressure 18000 psi (1240 Bar)
Particle Shape Spherical
Particle Size 1.7 μm
Pore Size 130 Å
Quantity 4/Pk.
Endcapped Yes
Type Method Development Kit
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.