missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation UPLC Method Development Scouting Kit
missing translation for 'orderingAttributeHoverText'
Length (Metric):
50 mm
100 mm
Diameter (Metric):
2.1 mm
3 mm
missing translation for 'unitSize'
Pack of 4
Description
- By utilizing UPLC Technology for method development, a substantial improvement in throughput can be realized.
- This, in turn, reduces cost per sample and time of analysis considerably while maintaining or improving separation integrity, enabling products to be brought to market faster and improving the overall profitability of the assay.
Specifications
Specifications
| Diameter (Metric) | 2.1 mm |
| Length (Metric) | 50 mm |
| Max. Pressure | 18000 psi (1240 Bar) |
| Particle Shape | Spherical |
| Particle Size | 1.7 μm |
| Pore Size | 130 Å |
| Quantity | 4/Pk. |
| Endcapped | Yes |
| Type | Method Development Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction