missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UPK3B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15709-1-AP-150UL
This item is not returnable.
View return policy
Description
Component of the asymmetric unit membrane (AUM); a highly specialized biomembrane elaborated by terminally differentiated urothelial cells. May play an important role in AUM-cytoskeleton interaction in terminally differentiated urothelial cells. It also contributes to the formation of urothelial glycocalyx which may play an important role in preventing bacterial adherence.Specifications
| UPK3B | |
| Polyclonal | |
| Unconjugated | |
| UPK3B | |
| P35, UP3b, UPIIIb, UPK3B, uroplakin 3B, Uroplakin IIIb | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100647, 105375355 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q80YF6, Q9BT76 | |
| UPK3B | |
| UPK3B Fusion Protein Ag8326 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction