missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UPK3A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553792
This item is not returnable.
View return policy
Description
Immunogen sequence: QILNAYLVRV GANGTCLWDP NFQGLCNPPL SAATEYRFKY VLVNMSTGLV EDQTLWSDPI RTNQLTPYST IDTWPGRRSG GMIVIT Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.
This gene encodes a member of the uroplakin family, a group of transmembrane proteins that form complexes on the apical surface of the bladder epithelium. Mutations in this gene may be associated with renal adysplasia. Alternatively spliced transcript variants have been described.
Specifications
| UPK3A | |
| Polyclonal | |
| Unconjugated | |
| Upk3a | |
| 1110017C07Rik; proplakin IIIa; UP3a; UPIII; UPIIIA; Upk3; UPK3A; uroplakin 3; uroplakin 3A; uroplakin III; Uroplakin-3a | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7380 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75631 | |
| Upk3a | |
| Recombinant protein corresponding to Human UPK3A. Recombinant protein control fragment (Product #RP-92355). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction