missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UPF2 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55022-1-AP-20UL
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
The UPF2 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human UPF2. This antibody recognizes human antigen. The UPF2 antibody has been validated for the following applications: WB, ELISA analysis.Tekniske data
| UPF2 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 26019 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, ELISA | |
| 260 μg/mL | |
| DKFZp434D222; HUPF2; KIAA1408; MGC138834; MGC138835; RENT2; smg-3 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| 148 kDa | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion