missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UPF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22281
This item is not returnable.
View return policy
Description
UPF2 Polyclonal specifically detects UPF2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| UPF2 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| DKFZP434D222, hUpf2, KIAA1408MGC138835, Nonsense mRNA reducing factor 2, regulator of nonsense transcripts 2, RENT2MGC138834, smg-3, UPF2 regulator of nonsense transcripts homolog (yeast), Up-frameshift suppressor 2 homolog, yeast Upf2p homolog | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 26019 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:1000 - 1:5000, Immunoprecipitation 2 - 10 ug/mg | |
| NP_056357.1 | |
| UPF2 | |
| The immunogen this antibody was made to, maps to a region between residue 1 and 50 of human Regulator of Nonsense Transcripts 2 using the numbering given in entry NP_056357.1 (GeneID 26019). | |
| 100 μL | |
| Primary | |
| Human, Mouse. Based on 100% sequence identity, this antibody is predicted to react with Bovine, Dog, Horse, Rabbit, Pig, Panda, Gorilla, Chimpanzee, Duckbill platypus, White-tufted-ear marmoset, European domestic ferret, Gray short-tailed opossum, Small-eared galago, Little brown bat and Northern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction