missing translation for 'onlineSavingsMsg'
Learn More

UPF2 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18401226 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401226 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401226 Supplier Novus Biologicals Supplier No. NBP222281UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UPF2 Polyclonal antibody specifically detects UPF2 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen UPF2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DKFZP434D222, hUpf2, KIAA1408MGC138835, Nonsense mRNA reducing factor 2, regulator of nonsense transcripts 2, RENT2MGC138834, smg-3, UPF2 regulator of nonsense transcripts homolog (yeast), Up-frameshift suppressor 2 homolog, yeast Upf2p homolog
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1 and 50 of human Regulator of Nonsense Transcripts 2 using the numbering given in entry NP_056357.1 (GeneID 26019).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26019
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.