Learn More
UPF1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28384-1-AP-20UL
 View more versions of this product
Product Code. 16826125
  • £135.00 / 20µL
Description
The UPF1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human UPF1. This antibody recognizes human antigen. The UPF1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
UPF1
Polyclonal
Unconjugated
20 μL
5976
IgG
Western Blot, Immunohistochemistry, ELISA
267 μg/mL