missing translation for 'onlineSavingsMsg'
Learn More

UPF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£111.18 - £355.98

Specifications

Antigen UPF1
Concentration 0.33 mg/mL
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16862325
View Documents
Proteintech
23379-1-AP-20UL
20 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16852325
View Documents
Proteintech
23379-1-AP-150UL
150 μL
£355.98
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Upf1 was identified as an active component in nonsense-mediated decay (NMD), an mRNA surveillance mechanism in eukaryotic cells that degrades mRNAs containing premature termination codons. Upf1 was found to be an ATP-dependent RNA helicase in the cytoplasm and was later shown to be a component of cytoplasmic P-bodies. Upf1 phosphorylation mediates the repression of translation that accompanies NMD, allowing mRNA accessibility to the NMD machinery. Two other active components of NMD, Upf2 and Upf3, were also identified and described as having perinuclear and nucleocytoplasmic localization, respectively.
Specifications

Specifications

UPF1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Rat, Mouse, Human
Q92900, Q9EPU0
19704, 5976, 684558
UPF1 Fusion Protein Ag16195
Primary
-20°C
UPF1
0.33 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
ATP dependent helicase RENT1, HUPF1, KIAA0221, NORF1, pNORF1, RENT1, UPF1
UPF1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.