missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UPF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554031
This item is not returnable.
View return policy
Description
Immunogen sequence: LEELWKENPS ATLEDLEKPG VDEEPQHVLL RYEDAYQYQN IFGPLVKLEA DYDKKLKESQ TQDNITVRWD LGLNKKRIAY FTLPKTD Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
Upf1 was identified as an active component in nonsense-mediated decay (NMD), an mRNA surveillance mechanism in eukaryotic cells that degrades mRNAs containing premature termination codons. Upf1 was found to be an ATP-dependent RNA helicase in the cytoplasm and was later shown to be a component of cytoplasmic P-bodies. Upf1 phosphorylation mediates the repression of translation that accompanies NMD, allowing mRNA accessibility to the NMD machinery. Two other active components of NMD, Upf2 and Upf3, were also identified and described as having perinuclear and nucleocytoplasmic localization, respectively.
Specifications
| UPF1 | |
| Polyclonal | |
| Unconjugated | |
| UPF1 | |
| ATP-dependent helicase RENT1; B430202H16Rik; delta helicase; FLJ43809; FLJ46894; HUPF1; KIAA0221; LOW QUALITY PROTEIN: regulator of nonsense transcripts 1; mUpf1; Nonsense mRNA reducing factor 1; NORF1; pNORF1; PNORF-1; Regulator of nonsense transcripts 1; Regulator of nonsense transcripts 1 (Nonsense mRNA reducing factor 1) (NORF1) (Up-frameshift suppressor 1 homolog); Rent1; smg-2; smg-2 homolog, nonsense mediated mRNA decay factor; Upf1; UPF1 regulator of nonsense transcripts homolog; UPF1 regulator of nonsense transcripts homolog (yeast); UPF1 RNA helicase and ATPase; UPF1, RNA helicase and ATPase; Upflp; up-frameshift mutation 1 homolog; up-frameshift suppressor 1 homolog; wu:fi40f07; wu:fj48a01; yeast Upf1p homolog; zgc:55472 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5976 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92900 | |
| UPF1 | |
| Recombinant protein corresponding to Human UPF1. Recombinant protein control fragment (Product #RP-91824). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction