missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UPF1 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5149082

Product Code. 30536238

  • £357.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

Upf1 was identified as an active component in nonsense-mediated decay (NMD), an mRNA surveillance mechanism in eukaryotic cells that degrades mRNAs containing premature termination codons. Upf1 was found to be an ATP-dependent RNA helicase in the cytoplasm and was later shown to be a component of cytoplasmic P-bodies. Upf1 phosphorylation mediates the repression of translation that accompanies NMD, allowing mRNA accessibility to the NMD machinery. Two other active components of NMD, Upf2 and Upf3, were also identified and described as having perinuclear and nucleocytoplasmic localization, respectively.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

UPF1
Polyclonal
Unconjugated
UPF1
ATP-dependent helicase RENT1; B430202H16Rik; delta helicase; FLJ43809; FLJ46894; HUPF1; KIAA0221; LOW QUALITY PROTEIN: regulator of nonsense transcripts 1; mUpf1; Nonsense mRNA reducing factor 1; NORF1; pNORF1; PNORF-1; Regulator of nonsense transcripts 1; Regulator of nonsense transcripts 1 (Nonsense mRNA reducing factor 1) (NORF1) (Up-frameshift suppressor 1 homolog); Rent1; smg-2; smg-2 homolog, nonsense mediated mRNA decay factor; Upf1; UPF1 regulator of nonsense transcripts homolog; UPF1 regulator of nonsense transcripts homolog (yeast); UPF1 RNA helicase and ATPase; UPF1, RNA helicase and ATPase; Upflp; up-frameshift mutation 1 homolog; up-frameshift suppressor 1 homolog; wu:fi40f07; wu:fj48a01; yeast Upf1p homolog; zgc:55472
Goat
Antigen affinity chromatography
RUO
5976
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
Q92900
UPF1
E. coli-derived recombinant human RENT1/UPF1, Asp351-Arg600.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.