missing translation for 'onlineSavingsMsg'
Learn More

UPF 1069, MedChemExpress

Product Code. p-200100532
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16476078

Brand: Medchem Express HY14478

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress UPF 1069 is a PARP inhibitor, with IC50s of 8 and 0.3 μM for PARP-1 and PARP-2, respectively.

UPF 1069 is a PARP inhibitor, with IC50s of 8 and 0.3 μM for PARP-1 and PARP-2, respectively.

Specifications

Chemical Name or Material UPF 1069
CAS 1048371-03-4
Color White
Purity Grade Notes Research
Molecular Formula C17H13NO3
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (358.05 mM)
SMILES O=C1NC=CC2=C1C=CC=C2OCC(C3=CC=CC=C3)=O
Molecular Weight (g/mol) 279.29
Formula Weight 279.29
Percent Purity 99.2%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.