Learn More
Tocris Bioscience™ UPF 1069
PARP-2 inhibitor
Specifications
Purity >99%
View More Specs
Description
Description
Selective poly(ADP-ribose) polymerase (PARP) 2 inhibitor (IC50 values are 0.3 and 8.0 μM for PARP-2 and PARP-1 respectively).
Specifications
Specifications
>99%