Learn More
Idex Stainless-Steel Tubing 1/16 in. OD with Blue Band
Seamless, precut stainless steel tubing is specially prepared for the exacting requirements of todays analyses
Brand:  Idex U133
Encompass
Product Code. 12903520
Description
Description
Stainless steel is naturally self-passivating, forming an oxidation layer on newly created surfaces. Idex Heath & Science Scientific takes extra steps to ensure the chemical resistance of the stainless steel tubing by manually passivating before and after the tubing is cut into specified lengths.
  • Precut 316 stainless steel tubing:
  • Ends are machine cut and polished
  • Edges are deburred inside and outside
  • Tubing is passivated
  • Inside is flushed with reagent-grade isopropanol
  • Color-coded banding for easy identification of the inner diameter
  • All stainless-steel tubing longer than 1m is coiled
Specifications
Specifications
Tubing
1/16 in.
0.25 mm
1 m
19,700 psi (1358 bar)
Blue
0.010 in.
3.2 ft.
Stainless Steel (316)