Learn More
Idex Low-Pressure Inline Filters
These filters offer excellent protection and allow for tailoring to a variety of low-pressure applications.
Brand:  Idex 42718-36
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12973210
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Each filter is rated to 1,000 psi (68.9 bar). They work best when used with PTFE or ETFE tubing, but can be used with any appropriate size semi-rigid or rigid tubing.
  • Zero-dead volume connections
  • No wrenches required
  • For 1/16 in. or 1/8 in. OD tubing
When choosing alternate or replacement frits, select only those with an overall diameter of .200 in., including polymer ring.
Specifications
Specifications
2 μm
1/16 in. O.D. tubing
Stainless Steel