missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Idex High-Pressure Solvent Inline Filters
Description
In-line Solvent Filters for running cleaner systems
- Help prevent particulates from clogging equipment
- Parts made of stainless steel and PEEK
- Replacement frits available
- Max pressure 6000 psi (414 bar)
Specifications
Specifications
| Pore Size | 0.5 μm |
| Housing Material | PEEK™ |
| Type | High-Pressure Inline Filter |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?