missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UPB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121510
This item is not returnable.
View return policy
Description
UPB1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
UPB1 is a protein that belongs to the CN hydrolase family. Beta-ureidopropionase catalyzes the last step in the pyrimidine degradation pathway. The pyrimidine bases uracil and thymine are degraded via the consecutive action of dihydropyrimidine dehydrogenase (DHPDH), dihydropyrimidinase (DHP) and beta-ureidopropionase (UP) to beta-alanine and beta-aminoisobutyric acid, respectively. UP deficiencies are associated with N-carbamyl-beta-amino aciduria and may lead to abnormalities in neurological activity. This gene encodes a protein that belongs to the CN hydrolase family. Beta-ureidopropionase catalyzes the last step in the pyrimidine degradation pathway. The pyrimidine bases uracil and thymine are degraded via the consecutive action of dihydropyrimidine dehydrogenase (DHPDH), dihydropyrimidinase (DHP) and beta-ureidopropionase (UP) to beta-alanine and beta-aminoisobutyric acid, respectively. UP deficiencies are associated with N-carbamyl-beta-amino aciduria and may lead to abnormalities in neurological activity.
Specifications
| UPB1 | |
| Polyclonal | |
| Unconjugated | |
| UPB1 | |
| AI195023; Beta-alanine synthase; beta-ureidopropionase; beta-ureidopropionase 1; BUP1; BUP-1; N-carbamoyl-beta-alanine amidohydrolase; UPB1; ureidopropionase, beta; wu:fb69e03; zgc:64020 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100155919, 116593, 51733 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q03248, Q9UBR1 | |
| UPB1 | |
| Recombinant protein Beta-ureidopropionase. The antigen corresponds to amino acid range 121-384 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Pig, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction