Learn More
uPAR Antibody (ATN-658) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-28067JF525
Product Code. 30168870
  • £319.00 / 0.10mL
Description
uPAR Monoclonal antibody specifically detects uPAR in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
uPAR
Monoclonal
Janelia Fluor 525
CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3
PLAUR/ uPAR/ CD87
0.1 mL
Cancer
5329
Store at 4°C in the dark.
IgG1
Flow Cytometry, ELISA, Functional Assay
ATN-658
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified