missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uPAR Antibody (A6E8-R), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-33084
This item is not returnable.
View return policy
Description
uPAR Monoclonal antibody specifically detects uPAR in Human samples. It is validated for Western Blot
Specifications
| uPAR | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
| Recombinant protein within human uPAR aa 51-250/335. (Uniprot: Q03405) | |
| 100 μL | |
| Cancer | |
| 5329 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| A6E8-R | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction