Learn More
uPAR Antibody (109801), mFluor Violet 500 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB531MFV500
This item is not returnable.
View return policy
Description
uPAR Monoclonal antibody specifically detects uPAR in Mouse samples. It is validated for Western Blot, Flow Cytometry, Blocking Assay, CyTOF-reported
Specifications
| uPAR | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Cancer | |
| 5329 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, Blocking Assay, CyTOF | |
| 109801 | |
| Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-reported | |
| CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse uPAR isoform 1, Leu24-Thr297, Accession # P35456 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG2a |